missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl methylenediphosphonate, 97%

Catalog No. AC308820050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC308820050 5 mL Glass bottle
AC308820250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC308820050 Supplier Thermo Scientific Chemicals Supplier No. 308820050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1660-94-2 | C9H22O6P2 | 288.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 1660-94-2
Molecular Formula C9H22O6P2
Molecular Weight (g/mol) 288.22
MDL Number MFCD00039887
InChI Key STJWVOQLJPNAQL-UHFFFAOYSA-N
Synonym tetraethyl methylenediphosphonate, tetraethyl methanediphosphonate, tetraethyl diphosphonomethane, methylenebis diethylphosphonate, tetraethyl methylenebisphosphonate, phosphonic acid, methylenebis-, tetraethyl ester, methylenebis diethoxyphosphine oxide, methylenebisphosphonic acid, tetraethyl ester, phosphonic acid, methylenedi-, tetraethyl ester, methylenedi phosphonic acid tetraethyl ester
PubChem CID 15455
IUPAC Name 1-[diethoxyphosphorylmethyl(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(CP(=O)(OCC)OCC)OCC

Specifications

Color Brown-Yellow to Yellow
Density 1.1600g/mL
Boiling Point 171.0°C to 174.0°C (11.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4405 to 1.4445
Linear Formula CH2[P(O)(OC2H5)2]2
Quantity 5 mL
Specific Gravity 1.16
Formula Weight 288.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tetraethyl methylenediphosphonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-764-5

RUO – Research Use Only