missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl Ethylenediphosphonate 98.0+%, TCI America™

Catalog No. T22945G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T22945G Supplier TCI America Supplier No. T22945G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 995-32-4
Molecular Formula C10H24O6P2
Molecular Weight (g/mol) 302.24
MDL Number MFCD00015133
InChI Key RSQYXXACEZCDFS-UHFFFAOYSA-N
Synonym tetraethyl ethylenediphosphonate, tetraethyl ethylenebisphosphonate, ethylenediphosphonic acid tetraethyl ester, tetraethyl ethane-1,2-diylbis phosphonate, 1,2-bis diethoxyphosphoryl ethane, 1,2-bis diethyl-phosphonato ethane, phosphonic acid, 1,2-ethanediylbis-, tetraethyl ester, diethyl 2-diethoxyphosphoryl ethylphosphonate, phosphonic acid, ethylenedi-, tetraethyl ester, acmc-209scy
PubChem CID 70446
IUPAC Name diethyl [2-(diethoxyphosphoryl)ethyl]phosphonate
SMILES CCOP(=O)(CCP(=O)(OCC)OCC)OCC

Specifications

Color Colorless
Boiling Point 202°C
Quantity 5 g
Formula Weight 302.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraethyl Ethylenediphosphonate