missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl 4-tert-Butylcalix[4]arene-O,O',O″,O‴-tetraacetate 96.0+%, TCI America™

Supplier:  TCI America T25631G

 View more versions of this product

Catalog No. T25631G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetraethyl 4-tert-Butylcalix[4]arene-O,O′,O′′,O′′′-tetraacetate
155°C
C60H80O12
1 g
HZHADWCIBZZJNV-UHFFFAOYSA-N
993.288
993.29
Crystalline Powder
97600-39-0
White-Yellow
MFCD00145373
sodium ionophore x, 4-tert-butylcalix 4 arene-tetraacetic acid tetraethyl ester, tetraethyl 4-tert-butylcalix 4 arene-o,o',o,o'-tetraacetate, p-tert-butylcalix 4 arene tetrakis ethyl acetate, sodium ionophore x, selectophore tm , function tested, 4-t-butylcalix 4 arene-tetraacetic acid tetraethyl ester, 4-tert-butylcalix 4 arene-tetraacetic acid tetraet, 4-tert-butylcalix 4 arenetetraacetic acid tetraethyl ester, 4-tert-butylcalix 4 arene-o,o',o,o'-tetraacetic acid tetraethyl ester
CCOC(=O)COC1=C2CC3=CC(=CC(=C3OCC(=O)OCC)CC4=C(C(=CC(=C4)C(C)(C)C)CC5=C(C(=CC(=C5)C(C)(C)C)CC1=CC(=C2)C(C)(C)C)OCC(=O)OCC)OCC(=O)OCC)C(C)(C)C
2724889
≥96.0% (HPLC)

Chemical Identifiers

97600-39-0
993.288
HZHADWCIBZZJNV-UHFFFAOYSA-N
2724889
C60H80O12
MFCD00145373
sodium ionophore x, 4-tert-butylcalix 4 arene-tetraacetic acid tetraethyl ester, tetraethyl 4-tert-butylcalix 4 arene-o,o',o,o'-tetraacetate, p-tert-butylcalix 4 arene tetrakis ethyl acetate, sodium ionophore x, selectophore tm , function tested, 4-t-butylcalix 4 arene-tetraacetic acid tetraethyl ester, 4-tert-butylcalix 4 arene-tetraacetic acid tetraet, 4-tert-butylcalix 4 arenetetraacetic acid tetraethyl ester, 4-tert-butylcalix 4 arene-o,o',o,o'-tetraacetic acid tetraethyl ester
CCOC(=O)COC1=C2CC3=CC(=CC(=C3OCC(=O)OCC)CC4=C(C(=CC(=C4)C(C)(C)C)CC5=C(C(=CC(=C5)C(C)(C)C)CC1=CC(=C2)C(C)(C)C)OCC(=O)OCC)OCC(=O)OCC)C(C)(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions