missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethoxymethane 97.0+%, TCI America™
$86.64 - $254.11
Chemical Identifiers
| CAS | 78-09-1 |
|---|---|
| Molecular Formula | C9H20O4 |
| Molecular Weight (g/mol) | 192.26 |
| MDL Number | MFCD00009221 |
| InChI Key | CWLNAJYDRSIKJS-UHFFFAOYSA-N |
| Synonym | tetraethyl orthocarbonate, tetraethoxymethane, triethoxymethoxy ethane, tetraethylorthocarbonate, ethane, 1,1',1,1'-methanetetrayltetrakis oxy tetrakis, orthocarbonic acid, tetraethyl ester, tetraethyi orthocarbonate, tetra ethyl orthocarbonate, 1-triethoxymethoxy ethane |
| PubChem CID | 66213 |
| IUPAC Name | (triethoxymethoxy)ethane |
| SMILES | CCOC(OCC)(OCC)OCC |
Chemical Identifiers
| 78-09-1 | |
| 192.26 | |
| CWLNAJYDRSIKJS-UHFFFAOYSA-N | |
| 66213 | |
| CCOC(OCC)(OCC)OCC |
| C9H20O4 | |
| MFCD00009221 | |
| tetraethyl orthocarbonate, tetraethoxymethane, triethoxymethoxy ethane, tetraethylorthocarbonate, ethane, 1,1',1,1'-methanetetrayltetrakis oxy tetrakis, orthocarbonic acid, tetraethyl ester, tetraethyi orthocarbonate, tetra ethyl orthocarbonate, 1-triethoxymethoxy ethane | |
| (triethoxymethoxy)ethane |
Specifications
| 78-09-1 | |
| 160°C | |
| MFCD00009221 | |
| 3271 | |
| CWLNAJYDRSIKJS-UHFFFAOYSA-N | |
| (triethoxymethoxy)ethane | |
| 66213 | |
| ≥97.0% (GC) | |
| Tetraethoxymethane |
| Colorless | |
| C9H20O4 | |
| 5 mL | |
| tetraethyl orthocarbonate, tetraethoxymethane, triethoxymethoxy ethane, tetraethylorthocarbonate, ethane, 1,1',1,1'-methanetetrayltetrakis oxy tetrakis, orthocarbonic acid, tetraethyl ester, tetraethyi orthocarbonate, tetra ethyl orthocarbonate, 1-triethoxymethoxy ethane | |
| CCOC(OCC)(OCC)OCC | |
| 192.26 | |
| 192.26 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator