missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethoxymethane 97.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 160°C |
| Quantity | 5 mL |
| UN Number | 3271 |
| Formula Weight | 192.26 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Tetraethoxymethane |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |