missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecylbenzene 97.0+%, TCI America™
Supplier: TCI America T086625G
Specifications
| Tetradecylbenzene | |
| 16°C | |
| 207°C | |
| MFCD00015080 | |
| 1-phenyltetradecane, benzene, tetradecyl, 1-tetradecylbenzene, tetradecane, 1-phenyl, myristylbenzene, n-tetradecylbenzene, unii-i0y9j0o95a, phenyl-n-tetradecane, acmc-209cvz, 1-phenyltetradecane, analytical standard | |
| CCCCCCCCCCCCCCC1=CC=CC=C1 | |
| 274.492 | |
| 274.49 | |
| Liquid |
| 1459-10-5 | |
| Yellow | |
| C20H34 | |
| 25 g | |
| JZALLXAUNPOCEU-UHFFFAOYSA-N | |
| tetradecylbenzene | |
| 15086 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 1459-10-5 | |
| 274.492 | |
| JZALLXAUNPOCEU-UHFFFAOYSA-N | |
| 15086 | |
| CCCCCCCCCCCCCCC1=CC=CC=C1 |
| C20H34 | |
| MFCD00015080 | |
| 1-phenyltetradecane, benzene, tetradecyl, 1-tetradecylbenzene, tetradecane, 1-phenyl, myristylbenzene, n-tetradecylbenzene, unii-i0y9j0o95a, phenyl-n-tetradecane, acmc-209cvz, 1-phenyltetradecane, analytical standard | |
| tetradecylbenzene |
Safety and Handling
EINECSNumber : (3)-0021
TSCA : Yes