missing translation for 'onlineSavingsMsg'
Learn More

Tetradecyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. T2265500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2265500ML 500 mL
T226525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2265500ML Supplier TCI America Supplier No. T2265500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21643-42-5
Molecular Formula C17H32O2
Molecular Weight (g/mol) 268.44
MDL Number MFCD00673244
InChI Key XZHNPVKXBNDGJD-UHFFFAOYSA-N
Synonym Acrylic Acid Tetradecyl Ester
PubChem CID 88984
IUPAC Name tetradecyl prop-2-enoate
SMILES CCCCCCCCCCCCCCOC(=O)C=C

Specifications

Melting Point 14°C
Color Yellow
Boiling Point 138°C
Quantity 500 mL
Formula Weight 268.44
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetradecyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0990
TSCA Yes