missing translation for 'onlineSavingsMsg'
Learn More

Tetradecanoic acid, 98%

Catalog No. AAA1206730
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1206730 250 g
AAA1206736 500 g
AAA120670E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1206730 Supplier Thermo Scientific Chemicals Supplier No. A1206730
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 544-63-8 | C14H28O2 | 228.38 g/mol

It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.

Chemical Identifiers

CAS 544-63-8
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
MDL Number MFCD00002744
InChI Key TUNFSRHWOTWDNC-UHFFFAOYSA-N
Synonym myristic acid, n-tetradecanoic acid, n-tetradecoic acid, crodacid, n-tetradecan-1-oic acid, 1-tridecanecarboxylic acid, hydrofol acid 1495, univol u 316s, emery 655, myristinsaeure
PubChem CID 11005
ChEBI CHEBI:28875
IUPAC Name tetradecanoic acid
SMILES CCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 53°C to 56°C
Density 0.862
Boiling Point 326°C
Flash Point 175°C (347°F)
Quantity 250 g
Beilstein 508624
Merck Index 14,6333
Solubility Information Insoluble in water.
Formula Weight 228.38
Percent Purity 98%
Chemical Name or Material Tetradecanoic acid
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 208-875-2
RTECSNumber QH4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only