missing translation for 'onlineSavingsMsg'
Learn More

Tetradecanedioic Acid 98.0+%, TCI America™

Catalog No. p-7136399
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07045G 5 g
T070425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07045G Supplier TCI America Supplier No. T07045G
Only null left

Chemical Identifiers

CAS 821-38-5
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.358
MDL Number MFCD00002743
InChI Key HQHCYKULIHKCEB-UHFFFAOYSA-N
Synonym 1,12-dodecanedicarboxylic acid, tetradecane-1,14-dioic acid, 1,14-tetradecanedioic acid, unii-uec3ac47h1, dodecamethylenedicarboxylic acid, tetradecanedicarboxylic acid, uec3ac47h1, 1,12-dodecanedicarboxylicacid, tetradecanedicarboxylate, pubchem18099
PubChem CID 13185
ChEBI CHEBI:76308
IUPAC Name tetradecanedioic acid
SMILES C(CCCCCCC(=O)O)CCCCCC(=O)O

Specifications

Melting Point 127°C
Color White
Quantity 5 g
Formula Weight 258.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetradecanedioic Acid
EINECSNumber (2)-0878
TSCA Yes