missing translation for 'onlineSavingsMsg'
Learn More

Tetradecanedioic acid, 98%

CAS: 821-38-5 | C14H26O4 | 258.358 g/mol

Supplier:  Thermo Scientific Chemicals H6025414

 View more versions of this product

Catalog No. AAH6025414


Only null left
Explore available promotions

Description

Description

Tetradecanedioic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetradecanedioic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents amnd bases.
Specifications

Specifications

Tetradecanedioic acid
124°C to 127°C
HOOC(CH2)12COOH
25 g
1,12-dodecanedicarboxylic acid, tetradecane-1,14-dioic acid, 1,14-tetradecanedioic acid, unii-uec3ac47h1, dodecamethylenedicarboxylic acid, tetradecanedicarboxylic acid, uec3ac47h1, 1,12-dodecanedicarboxylicacid, tetradecanedicarboxylate, pubchem18099
HQHCYKULIHKCEB-UHFFFAOYSA-N
tetradecanedioic acid
13185
258.35
821-38-5
C14H26O4
MFCD00002743
1788202
Slightly soluble in water.
C(CCCCCCC(=O)O)CCCCCC(=O)O
258.358
CHEBI:76308
98%

Chemical Identifiers

821-38-5
258.358
HQHCYKULIHKCEB-UHFFFAOYSA-N
13185
tetradecanedioic acid
C14H26O4
MFCD00002743
1,12-dodecanedicarboxylic acid, tetradecane-1,14-dioic acid, 1,14-tetradecanedioic acid, unii-uec3ac47h1, dodecamethylenedicarboxylic acid, tetradecanedicarboxylic acid, uec3ac47h1, 1,12-dodecanedicarboxylicacid, tetradecanedicarboxylate, pubchem18099
CHEBI:76308
C(CCCCCCC(=O)O)CCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 212-476-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only