missing translation for 'onlineSavingsMsg'
Learn More

Tetradecanedioic acid, 98%

Catalog No. AAH6025406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6025406 5 g
AAH6025414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6025406 Supplier Thermo Scientific Chemicals Supplier No. H6025406

CAS: 821-38-5 | C14H26O4 | 258.358 g/mol

Tetradecanedioic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetradecanedioic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents amnd bases.

Chemical Identifiers

CAS 821-38-5
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.358
MDL Number MFCD00002743
InChI Key HQHCYKULIHKCEB-UHFFFAOYSA-N
Synonym 1,12-dodecanedicarboxylic acid, tetradecane-1,14-dioic acid, 1,14-tetradecanedioic acid, unii-uec3ac47h1, dodecamethylenedicarboxylic acid, tetradecanedicarboxylic acid, uec3ac47h1, 1,12-dodecanedicarboxylicacid, tetradecanedicarboxylate, pubchem18099
PubChem CID 13185
ChEBI CHEBI:76308
IUPAC Name tetradecanedioic acid
SMILES C(CCCCCCC(=O)O)CCCCCC(=O)O

Specifications

Melting Point 124°C to 127°C
Linear Formula HOOC(CH2)12COOH
Quantity 5 g
Beilstein 1788202
Solubility Information Slightly soluble in water.
Formula Weight 258.35
Percent Purity 98%
Chemical Name or Material Tetradecanedioic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-476-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only