missing translation for 'onlineSavingsMsg'
Learn More

Tetradecane 99.0+%, TCI America™

Catalog No. T007925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T007925ML 25 mL
T0079500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T007925ML Supplier TCI America Supplier No. T007925ML

Chemical Identifiers

CAS 629-59-4
Molecular Formula C14H30
Molecular Weight (g/mol) 198.394
MDL Number MFCD00008986
InChI Key BGHCVCJVXZWKCC-UHFFFAOYSA-N
Synonym n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
PubChem CID 12389
ChEBI CHEBI:41253
IUPAC Name tetradecane
SMILES CCCCCCCCCCCCCC

Specifications

Melting Point 6°C
Color Colorless
Boiling Point 253°C
Quantity 25 mL
Formula Weight 198.39
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetradecane
EINECSNumber (2)-0010
RTECSNumber XB8000000
TSCA Yes