missing translation for 'onlineSavingsMsg'
Learn More

Tetradecane, (n-Tetradecane), Reagents

Catalog No. 01336934
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336934 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336934 Supplier Labchem Inc Supplier No. C2337530100C
Only null left

Tetradecane, (n-Tetradecane), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 629-59-4
Molecular Formula C14H30
Molecular Weight (g/mol) 198.39
InChI Key BGHCVCJVXZWKCC-UHFFFAOYSA-N
Synonym n-Tetradecane, Methyl tridecane
IUPAC Name tetradecane
SMILES CCCCCCCCCCCCCC

Specifications

Boiling Range 253°C
Melting Point 4°C
Color Colorless
Boiling Point 253°C
Melting Range 4°C
Packaging Glass Bottle
Quantity 100 mL
Specific Gravity 0.763
Grade Reagent
Physical Form Liquid
Chemical Name or Material Tetradecane, (n-Tetradecane)
Show More Show Less