missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecane, (n-Tetradecane), Reagents
Tetradecane, (n-Tetradecane), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2337530450C
Specifications
| Tetradecane, (n-Tetradecane) | |
| 253°C | |
| Colorless | |
| 4°C | |
| C14H30 | |
| 0.763 | |
| BGHCVCJVXZWKCC-UHFFFAOYSA-N | |
| tetradecane | |
| Reagent |
| 629-59-4 | |
| 4°C | |
| 253°C | |
| Glass Bottle | |
| 450 mL | |
| n-Tetradecane, Methyl tridecane | |
| CCCCCCCCCCCCCC | |
| 198.39 | |
| Liquid |
Chemical Identifiers
| 629-59-4 | |
| 198.39 | |
| n-Tetradecane, Methyl tridecane | |
| CCCCCCCCCCCCCC |
| C14H30 | |
| BGHCVCJVXZWKCC-UHFFFAOYSA-N | |
| tetradecane |