missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Tetradecane

Catalog No. 172456500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
172456500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 172456500G Supplier Sigma Aldrich Supplier No. 172456500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetradecane

Specifications

CAS 629-59-4
Melting Point 5.5°C (lit.)
Density 0.762 g/mL (at 20°C (literature))
Boiling Point 252°C to 254°C (lit.)
Molecular Formula C14H30
Linear Formula CH3(CH2)12 CH3
Refractive Index n20/D 1.429 (literature)
MDL Number MFCD00008986
Quantity 500 g
Molecular Weight (g/mol) 198.39
Percent Purity ≥99%
Show More Show Less