missing translation for 'onlineSavingsMsg'
Learn More

Tetradecamethylcycloheptasiloxane 95.0+%, TCI America™

Catalog No. T26781ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26781ML 1 mL
T26785ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26781ML Supplier TCI America Supplier No. T26781ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-50-6
Molecular Formula C14H42O7Si7
Molecular Weight (g/mol) 519.08
MDL Number MFCD30534308
InChI Key GSANOGQCVHBHIF-UHFFFAOYSA-N
PubChem CID 7874
IUPAC Name tetradecamethylcycloheptasiloxane
SMILES C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1

Specifications

Melting Point -32°C
Color Colorless
Boiling Point 151°C
Quantity 1 mL
Formula Weight 519.08
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetradecamethylcycloheptasiloxane