missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecafluoromethylcyclohexane 98.0+%, TCI America™
Supplier: TCI America P0846100G
Specifications
| Tetradecafluoromethylcyclohexane | |
| Colorless | |
| C7F14 | |
| 100 g | |
| QIROQPWSJUXOJC-UHFFFAOYSA-N | |
| 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-(trifluoromethyl)cyclohexane | |
| 9637 | |
| ≥98.0% (GC) |
| 355-02-2 | |
| 76°C | |
| MFCD00001468 | |
| perfluoro methylcyclohexane, perfluoromethylcyclohexane, flutec pp2, undecafluoro trifluoromethyl cyclohexane, tetradecafluoromethylcyclohexane, cyclohexane, undecafluoro trifluoromethyl, unii-vj9772yw63, 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-trifluoromethyl cyclohexane, cyclohexane, 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-trifluoromethyl, perfluoro methylcyclohexane , pmch | |
| FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)F | |
| 350.06 | |
| 350.05 | |
| Liquid |
Chemical Identifiers
| 355-02-2 | |
| 350.06 | |
| QIROQPWSJUXOJC-UHFFFAOYSA-N | |
| 9637 | |
| FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)F |
| C7F14 | |
| MFCD00001468 | |
| perfluoro methylcyclohexane, perfluoromethylcyclohexane, flutec pp2, undecafluoro trifluoromethyl cyclohexane, tetradecafluoromethylcyclohexane, cyclohexane, undecafluoro trifluoromethyl, unii-vj9772yw63, 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-trifluoromethyl cyclohexane, cyclohexane, 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-trifluoromethyl, perfluoro methylcyclohexane , pmch | |
| 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-(trifluoromethyl)cyclohexane |
Safety and Handling
RTECSNumber : GV7610000
TSCA : Yes