missing translation for 'onlineSavingsMsg'
Learn More

Tetradecafluoromethylcyclohexane 98.0+%, TCI America™

Catalog No. P084625G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P084625G 25 g
P0846100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P084625G Supplier TCI America Supplier No. P084625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 355-02-2
Molecular Formula C7F14
Molecular Weight (g/mol) 350.06
MDL Number MFCD00001468
InChI Key QIROQPWSJUXOJC-UHFFFAOYSA-N
Synonym perfluoro methylcyclohexane, perfluoromethylcyclohexane, flutec pp2, undecafluoro trifluoromethyl cyclohexane, tetradecafluoromethylcyclohexane, cyclohexane, undecafluoro trifluoromethyl, unii-vj9772yw63, 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-trifluoromethyl cyclohexane, cyclohexane, 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-trifluoromethyl, perfluoro methylcyclohexane , pmch
PubChem CID 9637
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-(trifluoromethyl)cyclohexane
SMILES FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)F

Specifications

Color Colorless
Boiling Point 76°C
Quantity 25 g
Formula Weight 350.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetradecafluoromethylcyclohexane
RTECSNumber GV7610000
TSCA Yes