missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecafluorohexane 96.0+%, TCI America™
$432.46 - $2736.43
Chemical Identifiers
| CAS | 355-42-0 |
|---|---|
| Molecular Formula | C6F14 |
| Molecular Weight (g/mol) | 338.044 |
| MDL Number | MFCD00000437 |
| InChI Key | ZJIJAJXFLBMLCK-UHFFFAOYSA-N |
| Synonym | perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn |
| PubChem CID | 9639 |
| ChEBI | CHEBI:39427 |
| IUPAC Name | 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane |
| SMILES | C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F |
Chemical Identifiers
| 355-42-0 | |
| 338.044 | |
| ZJIJAJXFLBMLCK-UHFFFAOYSA-N | |
| 9639 | |
| 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane |
| C6F14 | |
| MFCD00000437 | |
| perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn | |
| CHEBI:39427 | |
| C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F |
Specifications
| 355-42-0 | |
| 60°C | |
| MFCD00000437 | |
| perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn | |
| C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F | |
| 338.044 | |
| CHEBI:39427 | |
| ≥96.0% (GC) | |
| Tetradecafluorohexane |
| Colorless | |
| C6F14 | |
| 25 g | |
| ZJIJAJXFLBMLCK-UHFFFAOYSA-N | |
| 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane | |
| 9639 | |
| 338.04 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2366
RTECSNumber : MO4310000
TSCA : Yes