missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline Hydrochloride, Fisher BioReagents

Catalog No. BP912100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP912100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP912100 Supplier Fisher BioReagents Supplier No. BP912 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetracycline blocks protein synthesis and is used to select tetracycline-resistant recombinant plasmids.

Chemical Identifiers

CAS 64-75-5
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.898
InChI Key HTXDZWDXSWLLLW-FMZCEJRJSA-N
PubChem CID 129628373
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl

Specifications

Type Tetracycline Hydrochloride
Physical Form Solid
pH 2.5
Melting Point 223°C
Loss on Drying 2.0% max.
Packaging Poly Bottle
Potency > 900μg/mg
Specific Rotation -255 to -240°
Color Yellow
Quantity 100 g
Merck Index 15, 9341
Formula Weight 480.9
Chemical Name or Material Tetracycline Hydrochloride
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Substances known to cause developmental toxicity in humans. May cause skin and respiratory tract irritation. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. WARNING! This product contains a chemical known in the State of California to cause cancer.

NFPA

Health:2
Flammability:0
Instability:0
Recommended Storage RT