missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetracosanol, 99+%
CAS: 506-51-4 | C24H50O | 354.66 g/mol
Supplier: Thermo Scientific Chemicals 258150010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tetracosanol | |
| 75°C to 77°C | |
| 218°C | |
| 99+% | |
| C24H50O | |
| MFCD00042662 | |
| 1-tetracosanol, tetracosanol, lignoceryl alcohol, lignocerol, n-tetracosanol, tetracosyl alcohol, lignoceric alcohol, unii-2n0pi37ioc, n-tetracosanol-1, 2n0pi37ioc | |
| TYWMIZZBOVGFOV-UHFFFAOYSA-N | |
| tetracosan-1-ol | |
| 10472 | |
| 354.65 | |
| Shiny Flakes or Fluffy Powder |
| 506-51-4 | |
| White | |
| Authentic | |
| Glass bottle | |
| CH3(CH2)23OH | |
| 1 g | |
| Solubility in water: .. Other solubilities: soluble in hydrocarbons,alcohols,ether,ketones,,specially when hot | |
| CCCCCCCCCCCCCCCCCCCCCCCCO | |
| 354.66 | |
| CHEBI:77413 | |
| 99+% |
Chemical Identifiers
| 506-51-4 | |
| 354.66 | |
| TYWMIZZBOVGFOV-UHFFFAOYSA-N | |
| 10472 | |
| tetracosan-1-ol |
| C24H50O | |
| MFCD00042662 | |
| 1-tetracosanol, tetracosanol, lignoceryl alcohol, lignocerol, n-tetracosanol, tetracosyl alcohol, lignoceric alcohol, unii-2n0pi37ioc, n-tetracosanol-1, 2n0pi37ioc | |
| CHEBI:77413 | |
| CCCCCCCCCCCCCCCCCCCCCCCCO |
Safety and Handling
EINECSNumber : 208-043-9
RUO – Research Use Only