missing translation for 'onlineSavingsMsg'
Learn More

Tetracontane 97.0+%, TCI America™

Catalog No. p-7136288
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04491G 1 g
T04495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04491G Supplier TCI America Supplier No. T04491G
Only null left

Chemical Identifiers

CAS 4181-95-7
Molecular Formula C40H82
Molecular Weight (g/mol) 563.10
MDL Number MFCD00015267
InChI Key KUPLEGDPSCCPJI-UHFFFAOYSA-N
Synonym n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc
PubChem CID 20149
IUPAC Name tetracontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 82°C
Color White
Quantity 1 g
Formula Weight 563.10
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetracontane