missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetracontane, 98%
CAS: 4181-95-7 | C40H82 | 563.10 g/mol
$173.65 - $173.65
Chemical Identifiers
| CAS | 4181-95-7 |
|---|---|
| Molecular Formula | C40H82 |
| Molecular Weight (g/mol) | 563.10 |
| MDL Number | MFCD00015267 |
| InChI Key | KUPLEGDPSCCPJI-UHFFFAOYSA-N |
| Synonym | n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc |
| PubChem CID | 20149 |
| IUPAC Name | tetracontane |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC309165000
|
Thermo Scientific Chemicals
309165000 |
500 mg | Glass bottle |
Each for $173.65
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4181-95-7 | |
| 563.10 | |
| KUPLEGDPSCCPJI-UHFFFAOYSA-N | |
| 20149 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
| C40H82 | |
| MFCD00015267 | |
| n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc | |
| tetracontane |
Specifications
| 4181-95-7 | |
| White | |
| 97.5% min. (GC) | |
| C40H82 | |
| MFCD00015267 | |
| 01,III,592 | |
| Solubility in water: insoluble. Other solubilities: soluble in petroleum ether | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
| 563.10 | |
| 563.1 | |
| Powder or Flaky Solid |
| 80.0°C to 83.0°C | |
| Authentic | |
| Glass bottle | |
| CH3(CH2)38CH3 | |
| 500 mg | |
| n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc | |
| KUPLEGDPSCCPJI-UHFFFAOYSA-N | |
| tetracontane | |
| 20149 | |
| 98% | |
| Tetracontane |
Safety and Handling
EINECSNumber : 224-055-7
RUO – Research Use Only