missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorosilane 98.0+%, TCI America™

Catalog No. T0636500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0636500G 500 g
T063625G 25 g
T0636100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0636500G Supplier TCI America Supplier No. T0636500G

Chemical Identifiers

CAS 10026-04-7
Molecular Formula Cl4Si
Molecular Weight (g/mol) 169.89
MDL Number MFCD00011229
InChI Key FDNAPBUWERUEDA-UHFFFAOYSA-N
Synonym silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4
PubChem CID 24816
IUPAC Name tetrachlorosilane
SMILES Cl[Si](Cl)(Cl)Cl

Specifications

Boiling Point 59°C
Melting Point -68°C
Color Yellow
Physical Form Liquid
Quantity 500 g
UN Number 1818
Formula Weight 169.89
Percent Purity ≥98.0% (T)
Chemical Name or Material Tetrachlorosilane
EINECSNumber (1)-0258
RTECSNumber VW0525000
TSCA Yes