missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorophthalic anhydride, 98%

CAS: 117-08-8 | C8Cl4O3 | 285.885 g/mol

Supplier:  Thermo Scientific Chemicals A174040C

 View more versions of this product

Catalog No. AAA174040C

Only null left
Explore available promotions

Description

Description

Protection of amines as their tetrachlorophthalimide derivatives, by reaction with the anhydride in the presence of triethylamine, has been found to be useful in oligosaccharide chemistry. The group is stable to a wide range of reagents but can be selectively cleaved with ethylenediamine at lower temperatures than for the parent phthalimide derivatives; in particular, carboxylic esters are not cleaved under these conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Protection of amines as their tetrachlorophthalimide derivatives, by reaction with the anhydride in the presence of triethylamine, has been found to be useful in oligosaccharide chemistry. The group is stable to a wide range of reagents but can be selectively cleaved with ethylenediamine at lower temperatures than for the parent phthalimide derivatives; in particular, carboxylic esters are not cleaved under these conditions.

Solubility
Hydrolyzes slowly in water.

Notes
Hygroscopic. Store away from oxidizing agents, acids.
Specifications

Specifications

Tetrachlorophthalic anhydride
254°C to 258°C
362°C (683°F)
C8Cl4O3
10,000 g
211560
tetrachlorophthalic anhydride, tetrathal, 4,5,6,7-tetrachloro-1,3-isobenzofurandione, niagathal, 1,3-isobenzofurandione, 4,5,6,7-tetrachloro, phthalic anhydride, tetrachloro, 4,5,6,7-tetrachloroisobenzofuran-1,3-dione, 1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran, unii-76glw0lbek, ccris 6202
AUHHYELHRWCWEZ-UHFFFAOYSA-N
4,5,6,7-tetrachloro-2-benzofuran-1,3-dione
8326
285.9
117-08-8
349°C to 354°C
Weak
MFCD00005920
UN3077
Moisture sensitive
Hydrolyzes slowly in water.
C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O
285.885
CHEBI:59097
98%

Chemical Identifiers

117-08-8
285.885
AUHHYELHRWCWEZ-UHFFFAOYSA-N
8326
4,5,6,7-tetrachloro-2-benzofuran-1,3-dione
C8Cl4O3
MFCD00005920
tetrachlorophthalic anhydride, tetrathal, 4,5,6,7-tetrachloro-1,3-isobenzofurandione, niagathal, 1,3-isobenzofurandione, 4,5,6,7-tetrachloro, phthalic anhydride, tetrachloro, 4,5,6,7-tetrachloroisobenzofuran-1,3-dione, 1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran, unii-76glw0lbek, ccris 6202
CHEBI:59097
C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P305+P351+P338-P308+P313-P310-P333+P313-P342+P311-P363-P501c

H317-H318-H334-H350

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 204-171-4

RTECSNumber : TI3450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only