missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorocyclopropene, 98%

Catalog No. p-196417
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420190050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC420190050 Supplier Thermo Scientific Chemicals Supplier No. 420190050
Only null left

CAS: 6262-42-6 | C3Cl4 | 177.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6262-42-6
Molecular Formula C3Cl4
Molecular Weight (g/mol) 177.84
MDL Number MFCD00001267
InChI Key BLZOHTXDDOAASQ-UHFFFAOYSA-N
Synonym tetrachlorocyclopropene, cyclopropene, tetrachloro, 1,2,3,3-tetrachloro-cyclopropene, tetrachlorocycloprop-1-ene, acmc-20ajjz, dsstox_cid_1660, dsstox_rid_76272, dsstox_gsid_21660, cyclopropene,1,2,3,3-tetrachloro
PubChem CID 80428
IUPAC Name 1,2,3,3-tetrachlorocyclopropene
SMILES C1(=C(C1(Cl)Cl)Cl)Cl

Specifications

Color Colorless
Boiling Point 125°C to 130°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5063 to 1.5083
Quantity 5 g
Formula Weight 177.84
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetrachlorocyclopropene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 228-418-0

RUO – Research Use Only