Learn More
Tetrachloroauric(III) acid trihydrate, ACS reagent
Tetrachloroauric(III) Acid Trihydrate, >-49.0% (Au), HAuCl4.3H2O, CAS Number-16961-25-4, Gold trichloride Acid Trihydrate, Chloroauric Acid Trihydrate, 1g, Orange, 393.83, ACS Reagent, GHS Signal Word: Danger | CAS: 16961-25-4 | AuCl4H | 339.77 g/mol
Supplier: Thermo Scientific Chemicals 411070010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tetrachloroauric(III) Acid Trihydrate | |
| 16961-25-4 | |
| 1 g | |
| HAuCl4·3H2O | |
| Chloroauric Acid Trihydrate, Gold trichloride Acid Trihydrate | |
| VDLSFRRYNGEBEJ-UHFFFAOYSA-K | |
| trichlorogold hydrochloride | |
| 393.83 | |
| ACS Reagent |
| Orange | |
| Solid | |
| AuCl4H | |
| 15,212 | |
| Solubility in water: 150g/L (20°C). Other solubilities: soluble in alcohol and ether | |
| Cl.Cl[Au](Cl)Cl | |
| 339.77 | |
| ≥49.0% (Au) | |
| Glass Bottle |
Chemical Identifiers
| 16961-25-4 | |
| 339.77 | |
| trichlorogold hydrochloride |
| AuCl4H | |
| VDLSFRRYNGEBEJ-UHFFFAOYSA-K | |
| Cl.Cl[Au](Cl)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Danger
RUO – Research Use Only