missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroauric(III) Acid Trihydrate, 99.5%, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. p-4939046
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
EPS box
Fibre case
Catalog No. Quantity Packaging
M1015820001 1 g EPS box
M1015820005 5 g Fibre case
2 options

Catalog No. M1015820001

Supplier: Technidata 1.01582.0001

Only null left
For analysis

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
Synonym Gold(III)chloride hydrochloric acid, Gold trichloride hydrochloride
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Physical Form Solid
Quantity 1 g
Assay Percent Range 99.50%
UN Number UN 3260
Formula Weight 393.83
Odor Odorless
Packaging EPS box
Appearance Yellow solid
Chemical Name or Material Tetrachloroauric(III) Acid Trihydrate
Hazard Category H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 240-948-4