missing translation for 'onlineSavingsMsg'
Learn More

EMSURE™, MilliporeSigma™ Tetrachloroauric(III) Acid Trihydrate, 99.5%, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1015820001
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
EPS box
Fibre case
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1015820001 1 g EPS box
M1015820005 5 g Fibre case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1015820001 Supplier EMSURE™, MilliporeSigma™ Supplier No. 1.01582.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16961-25-4 | AuCl4H | 339.77 g/mol

For analysis

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
Synonym Gold(III)chloride hydrochloric acid, Gold trichloride hydrochloride
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Color Yellow
Physical Form Solid
Quantity 1 g
Assay Percent Range 99.50%
Linear Formula HAuCl4x3H2O
MDL Number MFCD00149904
UN Number UN 3260
Solubility Information Solubility in Water
InChI Formula InChI=1S/Au.4Cl/q-1;;;;/p+1
Formula Weight 393.83
Percent Purity 99.5%
Manufacturing Origin Germany
Identification Passes Test
Infrared Spectrum Conforms
Odor Odorless
Packaging EPS box
Product Line EMSURE
Appearance Yellow solid
Density 3.9 g/cm³
Chemical Name or Material Tetrachloroauric(III) Acid Trihydrate
Show More Show Less
Hazard Category H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 240-948-4