missing translation for 'onlineSavingsMsg'
Learn More

Gold Chloride Trihydrate, Certified ACS, Fisher Chemical™

Catalog No. p-31974
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
Glass Vial
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
G541 1 g Glass Vial
G545 5 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G541 Supplier Thermo Fisher Scientific Supplier No. G54 1
Only null left
  • Used as a gold catalyst or source of gold for organic reactions
  • Intended for research use only. Not for human or animal diagnostic purposes.

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149904
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
Synonym Gold(III) chloride trihydrate, Tetrachloroauric acid trihydrate, Hydrogren tetrachloroaurate(III), Tetrachloroauric(III) acid, Chloroauric acid trihydrate
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Melting Point 254°C
Color Yellow
Physical Form Solid
Quantity 1 g
Formula Weight 393.83
Percent Purity ≥49%
Grade Certified ACS
Identification Pass Test
Packaging Glass Vial
Density 3.9g/cm³
Chemical Name or Material Tetrachloroauric Acid Trihydrate
Show More Show Less
Recommended Storage Refrigerate