missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloro-p-benzoquinone, 99%

CAS: 118-75-2 | C6Cl4O2 | 245.86 g/mol

$263.80 - $263.80

Chemical Identifiers

CAS 118-75-2
Molecular Formula C6Cl4O2
Molecular Weight (g/mol) 245.86
MDL Number MFCD00001594
InChI Key UGNWTBMOAKPKBL-UHFFFAOYSA-N
Synonym chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone
PubChem CID 8371
ChEBI CHEBI:36703
IUPAC Name 2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione
SMILES ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC213561000
SDS View Documents
Thermo Scientific Chemicals
213561000
100 g Plastic bottle
Each for $263.80
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

118-75-2
245.86
UGNWTBMOAKPKBL-UHFFFAOYSA-N
8371
2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione
C6Cl4O2
MFCD00001594
chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone
CHEBI:36703
ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O

Specifications

118-75-2
>100°C
Authentic
Plastic bottle
C6Cl4(=O)2
100 g
01,125; 02,66; 03,46; 04,75
chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone
UGNWTBMOAKPKBL-UHFFFAOYSA-N
2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione
8371
245.88
Liquid
>290°C
0.5% max. (1 g, 105°C, 3 hrs)
98.5% min. (HPLC)
C6Cl4O2
MFCD00001594
07, 636
15, 2078
Solubility in water: insoluble. Other solubilities: soluble in ether,sparingly soluble in ccl4,cs2 and chloroform,practically insoluble in cold petroleum ether and,cold alcohol
ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O
245.86
CHEBI:36703
99%
Tetrachloro-p-benzoquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-274-4

RTECSNumber : DK6825000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only