missing translation for 'onlineSavingsMsg'
Learn More

Tetracaine, 98%

Catalog No. AC460820050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460820050 5 g
AC460820250 25 g
AC460821000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC460820050 Supplier Thermo Scientific Chemicals Supplier No. 460820050

CAS: 94-24-6 | C15H24N2O2 | 264.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94-24-6
Molecular Formula C15H24N2O2
Molecular Weight (g/mol) 264.36
InChI Key GKCBAIGFKIBETG-UHFFFAOYSA-N
PubChem CID 5411
ChEBI CHEBI:9468

Specifications

Melting Point 43°C
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 264.36
Percent Purity 98%
Chemical Name or Material Tetracaine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
EINECSNumber 202-316-6

RUO – Research Use Only