missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylphosphonium hydroxide, 40 wt.% solution in water

Catalog No. AC395031000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC395031000 100 mL Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC395031000 Supplier Thermo Scientific Chemicals Supplier No. 395031000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14518-69-5 | C16H37OP | 276.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)

Specifications

Chemical Name or Material Tetrabutylphosphonium hydroxide
CAS 14518-69-5, 7732-18-5
Melting Point 15.0°C
Color Colorless to Yellow
Density 0.9990g/mL
Packaging Plastic Bottle
Molecular Formula C16H37OP
MDL Number MFCD00068456
Quantity 100 mL
Specific Gravity 0.999
Synonym tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o
Solubility Information Solubility in water: soluble.
InChI Key DFQPZDGUFQJANM-UHFFFAOYSA-M
SMILES [OH-].CCCC[P+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylphosphanium;hydroxide
Molecular Weight (g/mol) 276.45
PubChem CID 84487
Formula Weight 276.44
Percent Purity 37.5 to 42.5%
Physical Form Liquid
Show More Show Less

RUO – Research Use Only