missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylphosphonium Hydroxide (40% in Water), TCI America™

Catalog No. T257125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T257125G 25 g
T2571250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T257125G Supplier TCI America Supplier No. T257125G

Chemical Identifiers

CAS 14518-69-5
Molecular Formula C16H37OP
Molecular Weight (g/mol) 276.45
MDL Number MFCD00068456
InChI Key DFQPZDGUFQJANM-UHFFFAOYSA-M
Synonym tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o
PubChem CID 84487
IUPAC Name tetrabutylphosphanium hydroxide
SMILES [OH-].CCCC[P+](CCCC)(CCCC)CCCC

Specifications

Color Yellow
Quantity 25 g
UN Number 3267
Formula Weight 276.44
Physical Form Liquid
Chemical Name or Material Tetrabutylphosphonium Hydroxide (40% in Water)