missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylphosphonium Bromide 99.0+%, TCI America™

Catalog No. T112425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T112425G 25 g
T1124100G 100 g
T1124500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T112425G Supplier TCI America Supplier No. T112425G

Chemical Identifiers

CAS 3115-68-2
Molecular Formula C16H36BrP
Molecular Weight (g/mol) 339.342
MDL Number MFCD00011853
InChI Key RKHXQBLJXBGEKF-UHFFFAOYSA-M
Synonym tetrabutylphosphonium bromide, tetra-n-butylphosphonium bromide, phosphonium, tetrabutyl-, bromide, px 4b, tetra butyl phosphonium bromide, tetrabutylphosphoniumbromide, tetrabutylphosphanium bromide, tetrabutyl phosphonium bromide, phosphonium, tetrabutyl-, bromide 1:1, acmc-1clh3
PubChem CID 76564
IUPAC Name tetrabutylphosphanium;bromide
SMILES CCCC[P+](CCCC)(CCCC)CCCC.[Br-]

Specifications

Melting Point 104°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 339.34
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylphosphonium Bromide
EINECSNumber (2)-3487
RTECSNumber TA2417000
TSCA Yes