missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylphosphonium Bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III) Complex 98.0+%, TCI America™

Catalog No. T14161G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14161G Supplier TCI America Supplier No. T14161G
Only null left

Chemical Identifiers

CAS 105029-70-7
Molecular Formula C22H36NiPS10
MDL Number MFCD00191494

Specifications

Color Black-Green
Quantity 1 g
Formula Weight 710.80
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylphosphonium Bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III) Complex