missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Triiodide 96.0+%, TCI America™

Catalog No. T12715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12715G 5 g
T127125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12715G Supplier TCI America Supplier No. T12715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13311-45-0
Molecular Formula C48H108I3N3
Molecular Weight (g/mol) 1108.126
MDL Number MFCD00059123
InChI Key MZYZRSVLEXUIRP-UHFFFAOYSA-K
PubChem CID 23500181
IUPAC Name tetrabutylazanium;triiodide
SMILES CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[I-].[I-].[I-]

Specifications

Melting Point 71°C
Color Black-Purple
Quantity 5 g
Formula Weight 623.18
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Triiodide
EINECSNumber (2)-0186
TSCA No