missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Tetraphenylborate 98.0+%, TCI America™

Numéro de catalogue. T14801G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T14801G 1 g
T14805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T14801G Fournisseur TCI America Code fournisseur T14801G
Il en reste null

Identifiants chimiques

CAS 15522-59-5
Molecular Formula C40H56BN
Molecular Weight (g/mol) 561.71
MDL Number MFCD00011749
InChI Key ZHCCBGAUZWZGQV-UHFFFAOYSA-N
Synonym tetrabutylammonium tetraphenylborate, tetra-n-butylammonium tetraphenylborate, tetrabutylammoniumtetraphenylborate, tetraphenylboron tetrabutylammonium, tetrabutylazanium; tetraphenylboranuide, tetra-n-butylammonium tetraphenylborate, 99+%, tetrabutylammonium ion tetraphenylborate
PubChem CID 3084234
IUPAC Name tetrabutylazanium; tetraphenylboranuide
SMILES CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Spécifications

Melting Point 235°C
Color White
Quantity 1 g
Formula Weight 561.71
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Tetraphenylborate