missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Tetrakis(trifluoromethyl)cuprate(III) 98.0+%, TCI America™
$192.00 - $576.00
Chemical Identifiers
| CAS | 152376-94-8 |
|---|---|
| Molecular Formula | C20H36CuF12N-3 |
| Molecular Weight (g/mol) | 582.042 |
| InChI Key | ONKMOBXWIMXGQS-UHFFFAOYSA-N |
| Synonym | Tetrabutylammonium Tetrakis(trifluoromethyl)copper(III) |
| PubChem CID | 124220112 |
| IUPAC Name | copper;tetrabutylazanium;trifluoromethane |
| SMILES | CCCC[N+](CCCC)(CCCC)CCCC.[C-](F)(F)F.[C-](F)(F)F.[C-](F)(F)F.[C-](F)(F)F.[Cu] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T3374200MG
|
TCI America
T3374200MG |
200 mg |
Each for $192.00
|
|
|||||
|
T33741G
|
TCI America
T33741G |
1 g |
Each for $576.00
|
|
|||||
Chemical Identifiers
| 152376-94-8 | |
| 582.042 | |
| Tetrabutylammonium Tetrakis(trifluoromethyl)copper(III) | |
| copper;tetrabutylazanium;trifluoromethane |
| C20H36CuF12N-3 | |
| ONKMOBXWIMXGQS-UHFFFAOYSA-N | |
| 124220112 | |
| CCCC[N+](CCCC)(CCCC)CCCC.[C-](F)(F)F.[C-](F)(F)F.[C-](F)(F)F.[C-](F)(F)F.[Cu] |
Specifications
| 152376-94-8 | |
| White | |
| 200 mg | |
| ONKMOBXWIMXGQS-UHFFFAOYSA-N | |
| copper;tetrabutylazanium;trifluoromethane | |
| 124220112 | |
| ≥98.0% (T) | |
| Tetrabutylammonium Tetrakis(trifluoromethyl)cuprate(III) |
| 142°C | |
| C20H36CuF12N-3 | |
| Tetrabutylammonium Tetrakis(trifluoromethyl)copper(III) | |
| CCCC[N+](CCCC)(CCCC)CCCC.[C-](F)(F)F.[C-](F)(F)F.[C-](F)(F)F.[C-](F)(F)F.[Cu] | |
| 582.042 | |
| 582.04 | |
| Crystalline Powder |
Safety and Handling
TSCA : No