missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium tetrafluoroborate, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013239
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013239 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013239 Supplier Merck Emd Millipore Supplier No. 868975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

Molecular Weight (g/mol) 561.69
MDL Number MFCD00011749
Synonym Tetraphenylboron tetrabutylammonium

Specifications

Melting Point 230°C to 236°C; 233°C to 237°C
CAS 15522-59-5
Linear Formula (CH3 CH2 CH2 CH2)4 N[B(C6H5)4 ]
Quantity 5 g
Solubility Information Acetonitrile: 0.1 g/mL, clear, colorless
Formula Weight 561.69
Percent Purity ≥99.0% (NT); ≥99.0%
Grade For electrochemical analysis
Physical Form Powder