missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Phosphate (White to Off-White Powder/HPLC Grade), Fisher Chemical™
Improve HPLC separation of acidic analytes by ion pairing.
$6205.47 - $6205.47
Chemical Identifiers
| CAS | 5574-97-0 |
|---|---|
| Molecular Formula | C16H38NO4P |
| Molecular Weight (g/mol) | 339.46 |
| MDL Number | MFCD00064526 |
| InChI Key | ARRNBPCNZJXHRJ-UHFFFAOYSA-M |
| PubChem CID | 2735142 |
| IUPAC Name | tetrabutylazanium dihydrogen phosphate |
| SMILES | OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O6117100
|
Thermo Fisher Scientific
O6117100 |
100 g |
Each for $6,205.47
Case of 6 Each for $37,232.82
|
|
|||||
Description
- High purity
- Application tested
- Dissolves easily
Chemical Identifiers
| 5574-97-0 | |
| 339.46 | |
| ARRNBPCNZJXHRJ-UHFFFAOYSA-M | |
| tetrabutylazanium dihydrogen phosphate |
| C16H38NO4P | |
| MFCD00064526 | |
| 2735142 | |
| OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC |
Specifications
| 5574-97-0 | |
| 148°C | |
| 4 to 5 | |
| C16H38NO4P | |
| MFCD00064526 | |
| ARRNBPCNZJXHRJ-UHFFFAOYSA-M | |
| tetrabutylazanium dihydrogen phosphate | |
| 2735142 | |
| ≥98.5% | |
| Powder or Solid |
| 0.02 AU max. at 214nm, 0.1 AU max. at 210nm, 0.2 AU max. at 200nm | |
| White | |
| ≥98.5 % | |
| (CH3CH2CH2CH2)4NH2PO4 | |
| 100 g | |
| OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC | |
| 339.46 | |
| 339.45 | |
| HPLC | |
| Tetrabutylammonium Phosphate |
Safety and Handling