missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Tetrabutylammonium Phosphate Monobasic Solution
Description
Anion used to probe the structure, stability and intramolecular flexibility of a series of azacrown ethers.
Specifications
Specifications
| CAS | 5574-97-0 |
| Density | 1.037 g/mL at 25°C |
| Molecular Formula | C16H38NO4P |
| Linear Formula | (CH3CH2CH2CH2)4N[OP(OH)2O] |
| MDL Number | MFCD00064526 |
| Quantity | 100 mL |
| Synonym | Tetrabutylammonium dihydrogen phosphate |
| Molecular Weight (g/mol) | 339.45 g/mol |