missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Tetrabutylammonium Phosphate Monobasic Solution

Catalog No. 268100100ML
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
268100100ML 100 mL
268100500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 268100100ML Supplier Sigma Aldrich Supplier No. 268100100ML

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Anion used to probe the structure, stability and intramolecular flexibility of a series of azacrown ethers.

Specifications

CAS 5574-97-0
Density 1.037 g/mL at 25°C
Molecular Formula C16H38NO4P
Linear Formula (CH3CH2CH2CH2)4N[OP(OH)2O]
MDL Number MFCD00064526
Quantity 100 mL
Synonym Tetrabutylammonium dihydrogen phosphate
Molecular Weight (g/mol) 339.45 g/mol