Learn More
Tetrabutylammonium phosphate monobasic, 99%, HPLC grade
CAS: 5574-97-0 | C16H38NO4P | 339.46 g/mol
Supplier: Thermo Scientific Chemicals 420160250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography, Phase transfer catalystSpecifications
| Tetrabutylammonium phosphate monobasic | |
| 5574-97-0 | |
| 148.0°C to 151.0°C | |
| Authentic | |
| Glass bottle | |
| (CH3(CH2)3)4NH2PO4 | |
| 25 g | |
| Solubility in water: soluble. | |
| OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC | |
| 339.46 | |
| 339.45 | |
| Powder |
| 99% | |
| (0.1% aq. soln.) (1 cm quartz cell), (0.1% aq. soln.) (1 cm quartz cell) at 0nm, 0.05 max. at 214nm, 0.1 max. at 210nm, 0.2 max. at 200nm | |
| Gray to Black | |
| 99% | |
| C16H38NO4P | |
| MFCD00064526 | |
| tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion | |
| ARRNBPCNZJXHRJ-UHFFFAOYSA-M | |
| dihydrogen phosphate;tetrabutylazanium | |
| 2735142 | |
| 99% |
Chemical Identifiers
| 5574-97-0 | |
| 339.46 | |
| ARRNBPCNZJXHRJ-UHFFFAOYSA-M | |
| 2735142 | |
| OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC |
| C16H38NO4P | |
| MFCD00064526 | |
| tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion | |
| dihydrogen phosphate;tetrabutylazanium |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 226-947-1
TSCA : TSCA
RUO – Research Use Only