missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Tetrabutylammonium Phosphate Monobasic

Catalog No. 8683350G
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8683350G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 8683350G Supplier Sigma Aldrich Supplier No. 8683350G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrabutylammonium Phosphate Monobasic

Specifications

CAS 5574-97-0
Melting Point 151°C to 154°C
Molecular Formula C16H38NO4P
Linear Formula (CH3CH2CH2CH2)4N[OP(OH)2O]
MDL Number MFCD00064526
Quantity 5 g
Synonym Tetrabutylammonium dihydrogen phosphate
Molecular Weight (g/mol) 339.45 g/mol