missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium perchlorate, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. p-7187812
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019627 10 g
111018318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019627 Supplier Merck Emd Millipore Supplier No. 8689310G

Tetrabutylammonium perchlorate (TBAP) is a quaternary ammonium salt, mainly used as electrolyte.

Chemical Identifiers

MDL Number MFCD00038722

Specifications

Melting Point 211°C to 215°C
CAS 1923-70-2
Linear Formula (CH3 CH2 CH2 CH2)4 N(ClO4)
Quantity 10 g
Solubility Information Acetonitrile: 0.1 g/mL, clear, colorless
Formula Weight 341.91
Percent Purity ≥99.0% (T); ≥99.0%
Grade For electrochemical analysis