missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium methanesulfonate, ≥97.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111015081
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015081 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015081 Supplier Merck Emd Millipore Supplier No. 8687710G

Chemical Identifiers

MDL Number MFCD00043226
Synonym Methanesulfonic acid tetrabutylammonium salt

Specifications

Melting Point 78°C to 80°C
CAS 65411-49-6
Linear Formula [CH3 CH2 CH2 CH2]4 N(CH3 SO3)
Quantity 10 g
Formula Weight 337.56
Percent Purity ≥97.0% (T)