missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Iodide 98.0+%, TCI America™

Catalog No. T005725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T005725G 25 g
T0057100G 100 g
T0057500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T005725G Supplier TCI America Supplier No. T005725G

Chemical Identifiers

CAS 311-28-4
Molecular Formula C16H36IN
Molecular Weight (g/mol) 369.38
MDL Number MFCD00011636
InChI Key DPKBAXPHAYBPRL-UHFFFAOYSA-M
Synonym tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide
PubChem CID 67553
IUPAC Name tetrabutylazanium iodide
SMILES [I-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 147°C
Color White
Quantity 25 g
Formula Weight 369.38
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Iodide
EINECSNumber (2)-0186
RTECSNumber BS5450000
TSCA Yes