missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Hexafluorophosphate 98.0+%, TCI America™
Supplier: TCI America T1279250G
Specifications
| Tetrabutylammonium Hexafluorophosphate | |
| 245°C | |
| C16H36F6NP | |
| 250 g | |
| tetrabutylammonium hexafluorophosphate, tetra-n-butylammonium hexafluorophosphate, tetrabutylammonium hexafluorophosphate v, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-1:1, acmc-1ahjo, n,n,n-tributyl-1-butanaminium hexafluorophosphate, ksc225q6r, tetrabutylammoniumhexafluorophosphate, tetrabutylazanium hexafluorophosphate | |
| F[P-](F)(F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC | |
| 387.44 | |
| 387.44 | |
| Crystalline Powder |
| 3109-63-5 | |
| White | |
| MFCD00011748 | |
| 1759 | |
| BKBKEFQIOUYLBC-UHFFFAOYSA-N | |
| hexafluoro-λâµ-phosphanuide; tetrabutylazanium | |
| 165075 | |
| ≥98.0% (T) |
Chemical Identifiers
| 3109-63-5 | |
| 387.44 | |
| BKBKEFQIOUYLBC-UHFFFAOYSA-N | |
| 165075 | |
| F[P-](F)(F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC |
| C16H36F6NP | |
| MFCD00011748 | |
| tetrabutylammonium hexafluorophosphate, tetra-n-butylammonium hexafluorophosphate, tetrabutylammonium hexafluorophosphate v, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-1:1, acmc-1ahjo, n,n,n-tributyl-1-butanaminium hexafluorophosphate, ksc225q6r, tetrabutylammoniumhexafluorophosphate, tetrabutylazanium hexafluorophosphate | |
| hexafluoro-λâµ-phosphanuide; tetrabutylazanium |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : Yes