missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. T127925G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T127925G 25 g
T1279250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T127925G Supplier TCI America Supplier No. T127925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3109-63-5
Molecular Formula C16H36F6NP
Molecular Weight (g/mol) 387.44
MDL Number MFCD00011748
InChI Key BKBKEFQIOUYLBC-UHFFFAOYSA-N
Synonym tetrabutylammonium hexafluorophosphate, tetra-n-butylammonium hexafluorophosphate, tetrabutylammonium hexafluorophosphate v, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-1:1, acmc-1ahjo, n,n,n-tributyl-1-butanaminium hexafluorophosphate, ksc225q6r, tetrabutylammoniumhexafluorophosphate, tetrabutylazanium hexafluorophosphate
PubChem CID 165075
IUPAC Name hexafluoro-λ⁵-phosphanuide; tetrabutylazanium
SMILES F[P-](F)(F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 245°C
Color White
Quantity 25 g
UN Number 1759
Formula Weight 387.44
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Hexafluorophosphate
EINECSNumber (2)-0186
TSCA Yes